Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTCACATCCTTTTTGGCCAGAG[A/G]TCAGGCAATGGCTGTATGCTTCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610849 | ||||||||||||||||||||
Literature Links: |
TTLL6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TTLL6 - tubulin tyrosine ligase like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130918.1 | 2890 | Missense Mutation | ACC,ATC | T,I 882 | NP_001124390.1 | |
NM_173623.3 | 2890 | Missense Mutation | ACC,ATC | T,I 575 | NP_775894.2 | |
XM_017024491.1 | 2890 | Missense Mutation | ACC,ATC | T,I 881 | XP_016879980.1 | |
XM_017024492.1 | 2890 | Missense Mutation | ACC,ATC | T,I 729 | XP_016879981.1 | |
XM_017024493.1 | 2890 | Missense Mutation | ACC,ATC | T,I 729 | XP_016879982.1 | |
XM_017024494.1 | 2890 | Missense Mutation | ACC,ATC | T,I 729 | XP_016879983.1 | |
XM_017024495.1 | 2890 | Missense Mutation | ACC,ATC | T,I 635 | XP_016879984.1 | |
XM_017024496.1 | 2890 | Missense Mutation | ACC,ATC | T,I 574 | XP_016879985.1 | |
XM_017024497.1 | 2890 | Intron | XP_016879986.1 |