Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGACCATTATATTTGTCCCATAAC[A/G]TAACTGTTCTGCACAGTTAACATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123610 MIM: 609356 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRYBA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRYBA1 - crystallin beta A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUFIP2 - NUFIP2, FMR1 interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020772.2 | 7670 | UTR 3 | NP_065823.1 | |||
XM_017024896.1 | 7670 | UTR 3 | XP_016880385.1 |