Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGACGATCTGCTCGAGTCACCCG[C/T]TCCTCAGCCAGGCTAAGCCAGAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609880 | ||||||||||||||||||||
Literature Links: |
KAT7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KAT7 - lysine acetyltransferase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199155.1 | 348 | Silent Mutation | CGC,CGT | R,R 44 | NP_001186084.1 | |
NM_001199156.1 | 348 | Silent Mutation | CGC,CGT | R,R 44 | NP_001186085.1 | |
NM_001199157.1 | 348 | Silent Mutation | CGC,CGT | R,R 44 | NP_001186086.1 | |
NM_001199158.1 | 348 | Silent Mutation | CGC,CGT | R,R 44 | NP_001186087.1 | |
NM_007067.4 | 348 | Silent Mutation | CGC,CGT | R,R 44 | NP_008998.1 | |
XM_011524235.2 | 348 | Intron | XP_011522537.1 |