Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACAAGCTTTACATCGGCAACCTC[A/G]ACGAGAGCGTGACCCCCGCGGACTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608288 | ||||||||||||||||||||
Literature Links: |
IGF2BP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGF2BP1 - insulin like growth factor 2 mRNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160423.1 | 362 | Missense Mutation | AAC,GAC | N,D 10 | NP_001153895.1 | |
NM_006546.3 | 362 | Missense Mutation | AAC,GAC | N,D 10 | NP_006537.3 | |
XM_005256955.3 | 362 | Intron | XP_005257012.2 | |||
XM_011524201.2 | 362 | Missense Mutation | AAC,GAC | N,D 10 | XP_011522503.1 | |
XM_017024022.1 | 362 | Missense Mutation | AAC,GAC | N,D 10 | XP_016879511.1 |