Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGTTTGGAAATACTCTTTGTCT[A/G]GAGACACTTCTGTGGGCTACTTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606504 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASC3 - cancer susceptibility candidate 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAPGEFL1 - Rap guanine nucleotide exchange factor like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303533.1 | 1800 | Intron | NP_001290462.1 | |||
NM_001303534.1 | 1800 | Intron | NP_001290463.1 | |||
NM_016339.4 | 1800 | Intron | NP_057423.1 | |||
XM_006721932.3 | 1800 | Intron | XP_006721995.1 | |||
XM_006721935.3 | 1800 | Intron | XP_006721998.1 | |||
XM_006721937.3 | 1800 | UTR 5 | XP_006722000.1 | |||
XM_006721938.3 | 1800 | Intron | XP_006722001.1 |