Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAACTGGCATCAGCCTCCCTCT[G/T]GGACACGGACAAAAGGTATTGCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608463 MIM: 601999 | ||||||||||||||||||||
Literature Links: |
AATF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AATF - apoptosis antagonizing transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012138.3 | 445 | Missense Mutation | TGG,TTG | W,L 65 | NP_036270.1 | |
XM_011524611.1 | 445 | Missense Mutation | TGG,TTG | W,L 65 | XP_011522913.1 |
LHX1 - LIM homeobox 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |