Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCGCCAAGCCCAGACATGCAGGA[C/G]CTGCTCCGGAGCGTGGAGAGGGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614218 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR22HG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR22HG - MIR22 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR81 - WD repeat domain 81 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163673.1 | 216 | Intron | NP_001157145.1 | |||
NM_001163809.1 | 216 | Missense Mutation | GAC,GAG | D,E 29 | NP_001157281.1 | |
NM_001163811.1 | 216 | Intron | NP_001157283.1 | |||
NM_152348.3 | 216 | Intron | NP_689561.2 | |||
XM_011523651.2 | 216 | Intron | XP_011521953.1 | |||
XM_017024184.1 | 216 | Missense Mutation | GAC,GAG | D,E 29 | XP_016879673.1 |