Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGCCACGAGTGACCCTGCTGCC[C/T]ACGCCCCACGTCAGCGGGCTGAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609046 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTBD17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTBD17 - BTB domain containing 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPR142 - G protein-coupled receptor 142 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181790.1 | 387 | Silent Mutation | CCC,CCT | P,P 129 | NP_861455.1 | |
XM_005257305.3 | 387 | Silent Mutation | CCC,CCT | P,P 41 | XP_005257362.1 |