Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCGGCCGCTCATGTTCCTTGGAC[C/T]GCATGTGCTGGATATATACCTGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600362 MIM: 600966 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLII PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLII - FLII, actin remodeling protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256264.1 | 3707 | Missense Mutation | CAG,CGG | Q,R 1221 | NP_001243193.1 | |
NM_001256265.1 | 3707 | Missense Mutation | CAG,CGG | Q,R 1177 | NP_001243194.1 | |
NM_002018.3 | 3707 | Missense Mutation | CAG,CGG | Q,R 1232 | NP_002009.1 | |
XM_005256555.3 | 3707 | Missense Mutation | CAG,CGG | Q,R 1259 | XP_005256612.1 | |
XM_005256556.3 | 3707 | Missense Mutation | CAG,CGG | Q,R 1258 | XP_005256613.1 | |
XM_005256558.2 | 3707 | Missense Mutation | CAG,CGG | Q,R 1231 | XP_005256615.1 |
LLGL1 - LLGL1, scribble cell polarity complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004140.3 | 3707 | Intron | NP_004131.3 | |||
XM_011523849.2 | 3707 | Intron | XP_011522151.1 | |||
XM_011523850.2 | 3707 | Intron | XP_011522152.1 | |||
XM_011523851.2 | 3707 | Intron | XP_011522153.1 | |||
XM_011523852.2 | 3707 | Intron | XP_011522154.1 | |||
XM_011523853.2 | 3707 | Intron | XP_011522155.1 | |||
XM_011523854.2 | 3707 | Intron | XP_011522156.1 | |||
XM_011523855.2 | 3707 | Intron | XP_011522157.1 | |||
XM_011523856.1 | 3707 | Intron | XP_011522158.1 |