Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGCCGACCAGCCGGAGGGTGTT[A/G]GGGTAGCTCTTGCGCATGCTCATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612683 | ||||||||||||||||||||
Literature Links: |
TEKT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TEKT3 - tektin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031898.2 | 2947 | Silent Mutation | CCC,CCT | P,P 481 | NP_114104.1 | |
XM_011523988.2 | 2947 | Silent Mutation | CCC,CCT | P,P 481 | XP_011522290.1 | |
XM_011523989.1 | 2947 | Silent Mutation | CCC,CCT | P,P 481 | XP_011522291.1 | |
XM_011523990.2 | 2947 | Silent Mutation | CCC,CCT | P,P 315 | XP_011522292.1 | |
XM_017024954.1 | 2947 | Silent Mutation | CCC,CCT | P,P 481 | XP_016880443.1 | |
XM_017024955.1 | 2947 | Silent Mutation | CCC,CCT | P,P 315 | XP_016880444.1 | |
XM_017024956.1 | 2947 | Intron | XP_016880445.1 |