Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGATGAGGCACGTAGACCTCCTTA[A/G]TGTAGCGGACTATCCCCTCTTCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610885 MIM: 611837 MIM: 608125 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EME1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EME1 - essential meiotic structure-specific endonuclease 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL27 - mitochondrial ribosomal protein L27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016504.2 | 330 | Missense Mutation | ACT,ATT | T,I 100 | NP_057588.1 |
XYLT2 - xylosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |