Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAGACTTGCCTTATAGACAGCA[C/T]TGGGGAGGAGGTTGTTCATGGTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610664 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0100 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0100 - KIAA0100 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014680.3 | 6000 | Missense Mutation | AAT,AGT | N,S 1966 | NP_055495.2 | |
XM_005258073.2 | 6000 | Missense Mutation | AAT,AGT | N,S 1965 | XP_005258130.1 | |
XM_017025423.1 | 6000 | Missense Mutation | AAT,AGT | N,S 1823 | XP_016880912.1 |
SGK494 - uncharacterized serine/threonine-protein kinase SgK494 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPAG5-AS1 - SPAG5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |