Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTGCCTGAAACTTTCCTCCTGG[G/T]CAGGGGACCACCTCAGTCTCTCCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194480 MIM: 616463 | ||||||||||||||||||||
Literature Links: |
LOC101928000 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101928000 - uncharacterized LOC101928000 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZFP3 - ZFP3 zinc finger protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF232 - zinc finger protein 232 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320952.1 | 960 | Missense Mutation | GAC,GCC | D,A 272 | NP_001307881.1 | |
NM_001320953.1 | 960 | Missense Mutation | GAC,GCC | D,A 254 | NP_001307882.1 | |
NM_001320954.1 | 960 | Missense Mutation | GAC,GCC | D,A 254 | NP_001307883.1 | |
NM_001320955.1 | 960 | Missense Mutation | GAC,GCC | D,A 253 | NP_001307884.1 | |
NM_014519.3 | 960 | Missense Mutation | GAC,GCC | D,A 281 | NP_055334.2 | |
XM_011524006.2 | 960 | Missense Mutation | GAC,GCC | D,A 233 | XP_011522308.1 | |
XM_017025021.1 | 960 | Missense Mutation | GAC,GCC | D,A 281 | XP_016880510.1 | |
XM_017025022.1 | 960 | Missense Mutation | GAC,GCC | D,A 205 | XP_016880511.1 | |
XM_017025023.1 | 960 | Missense Mutation | GAC,GCC | D,A 196 | XP_016880512.1 |