Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCCTCGTCGTTGTCTTCCTCCT[C/T]CTGCTGCTCAACATCCATCCCGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616599 | ||||||||||||||||||||
Literature Links: |
BORCS6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BORCS6 - BLOC-1 related complex subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017622.2 | 527 | Missense Mutation | AAG,GAG | K,E 141 | NP_060092.2 |
MIR4521 - microRNA 4521 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |