Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGCTCCACCCAGTTGACCGGGCA[A/G]CAGGTCCTTTCTGAGCCTGAGCGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 108360 | ||||||||||||||||||||
Literature Links: |
ASGR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASGR1 - asialoglycoprotein receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197216.2 | 766 | Silent Mutation | TGC,TGT | C,C 114 | NP_001184145.1 | |
NM_001671.4 | 766 | Silent Mutation | TGC,TGT | C,C 153 | NP_001662.1 | |
XM_011523861.2 | 766 | Silent Mutation | TGC,TGT | C,C 153 | XP_011522163.1 |