Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGCCCTCCATCATCCCCCCAGA[A/G]AGGGTCAGAGTATCCCTTTCTGCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM92 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM92 - transmembrane protein 92 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168215.1 | 431 | Silent Mutation | GAA,GAG | E,E 107 | NP_001161687.1 | |
NM_153229.2 | 431 | Silent Mutation | GAA,GAG | E,E 107 | NP_694961.2 | |
XM_006721730.3 | 431 | Silent Mutation | GAA,GAG | E,E 107 | XP_006721793.1 |
TMEM92-AS1 - TMEM92 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |