Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAAGTGGTGTGTGGGCATCACCA[G/T]GCTTGTAGAGGACACTGCAGAGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 189907 | ||||||||||||||||||||
Literature Links: |
HNF1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HNF1B - HNF1 homeobox B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000458.3 | 1646 | Silent Mutation | GCA,GCC | A,A 556 | NP_000449.1 | |
NM_001165923.3 | 1646 | Silent Mutation | GCA,GCC | A,A 530 | NP_001159395.1 | |
NM_001304286.1 | 1646 | Missense Mutation | ATG,CTG | M,L 426 | NP_001291215.1 | |
XM_011525160.1 | 1646 | Missense Mutation | ATG,CTG | M,L 517 | XP_011523462.1 | |
XM_011525161.1 | 1646 | Silent Mutation | GCA,GCC | A,A 491 | XP_011523463.1 | |
XM_011525162.2 | 1646 | Intron | XP_011523464.1 | |||
XM_011525163.2 | 1646 | Intron | XP_011523465.1 | |||
XM_011525164.1 | 1646 | Missense Mutation | ATG,CTG | M,L 491 | XP_011523466.1 |