Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATGAAGAGAGGTGTTAGATACT[A/G]TTGAATTAGGCTACCCGGGGCACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601254 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP2K6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP2K6 - mitogen-activated protein kinase kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002758.3 | Intron | NP_002749.2 | ||||
XM_005257515.2 | Intron | XP_005257572.1 | ||||
XM_005257516.2 | Intron | XP_005257573.1 | ||||
XM_006721975.3 | Intron | XP_006722038.1 | ||||
XM_011525025.2 | Intron | XP_011523327.1 | ||||
XM_011525026.2 | Intron | XP_011523328.1 | ||||
XM_011525027.2 | Intron | XP_011523329.1 |