Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAGTGATGGAGGGGCTGCAGTT[C/T]GGAAATGAGTGTCTGAACAAGATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610901 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHMP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHMP6 - charged multivesicular body protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024591.4 | 396 | Silent Mutation | TTC,TTT | F,F 106 | NP_078867.2 | |
XM_005257668.1 | 396 | Silent Mutation | TTC,TTT | F,F 106 | XP_005257725.1 |
LOC100129503 - uncharacterized LOC100129503 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |