Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCTGCGCGCCCCCGGGCACGAC[C/G]CCGGAGTCTCCGCGGGCGGCCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613799 MIM: 606800 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC40 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC40 - coiled-coil domain containing 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243342.1 | 51 | Intron | NP_001230271.1 | |||
NM_017950.3 | 51 | Intron | NP_060420.2 | |||
XM_005257492.4 | 51 | Intron | XP_005257549.1 | |||
XM_011524963.2 | 51 | Intron | XP_011523265.1 | |||
XM_011524964.2 | 51 | Intron | XP_011523266.1 | |||
XM_011524965.2 | 51 | Intron | XP_011523267.1 | |||
XM_017024807.1 | 51 | Intron | XP_016880296.1 |
GAA - glucosidase alpha, acid | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000152.4 | 51 | UTR 5 | NP_000143.2 | |||
NM_001079803.2 | 51 | UTR 5 | NP_001073271.1 | |||
NM_001079804.2 | 51 | UTR 5 | NP_001073272.1 | |||
XM_005257193.2 | 51 | Intron | XP_005257250.1 | |||
XM_005257194.4 | 51 | UTR 5 | XP_005257251.1 |
MIR1268B - microRNA 1268b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |