Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCCACACAGCTGCCTGGAACCCA[C/T]GACTCCCGAGAGCCCGTCTGCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603127 MIM: 179618 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAS7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAS7 - growth arrest specific 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130831.1 | 7563 | UTR 3 | NP_001124303.1 | |||
NM_003644.2 | 7563 | UTR 3 | NP_003635.2 | |||
NM_201432.1 | 7563 | UTR 3 | NP_958836.1 | |||
NM_201433.1 | 7563 | UTR 3 | NP_958839.1 | |||
XM_005256831.3 | 7563 | UTR 3 | XP_005256888.1 | |||
XM_005256832.3 | 7563 | UTR 3 | XP_005256889.1 | |||
XM_005256833.3 | 7563 | UTR 3 | XP_005256890.1 | |||
XM_005256834.3 | 7563 | UTR 3 | XP_005256891.1 | |||
XM_005256835.3 | 7563 | UTR 3 | XP_005256892.1 | |||
XM_011524044.2 | 7563 | UTR 3 | XP_011522346.1 | |||
XM_017025239.1 | 7563 | UTR 3 | XP_016880728.1 | |||
XM_017025240.1 | 7563 | UTR 3 | XP_016880729.1 | |||
XM_017025241.1 | 7563 | UTR 3 | XP_016880730.1 | |||
XM_017025242.1 | 7563 | UTR 3 | XP_016880731.1 |
RCVRN - recoverin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |