Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTTTGCTTTTCAAAAGCAATGT[C/T]TTTATTGAATATAAAAGTGTATGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606245 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371727 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105371727 - uncharacterized LOC105371727 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUZ12 - SUZ12 polycomb repressive complex 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321207.1 | Intron | NP_001308136.1 | ||||
NM_015355.3 | Intron | NP_056170.2 | ||||
XM_006721794.3 | Intron | XP_006721857.1 | ||||
XM_017024409.1 | Intron | XP_016879898.1 |