Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCAAGCCGAGTCCTCCAACTAC[C/T]CCTGCTTGGGTGAGTACAGCCAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114760 | ||||||||||||||||||||
Literature Links: |
CA4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CA4 - carbonic anhydrase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005257639.2 | 197 | Missense Mutation | CCC,TCC | P,S 35 | XP_005257696.1 | |
XM_011525183.2 | 197 | UTR 5 | XP_011523485.1 | |||
XM_017025012.1 | 197 | UTR 5 | XP_016880501.1 |