Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAATATGTACTAAAGGTCTTGAA[G/T]CACCACTACAGCAAAAAAGGGCAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616934 | ||||||||||||||||||||
Literature Links: |
MEIOC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MEIOC - meiosis specific with coiled-coil domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145080.2 | 1101 | Missense Mutation | GCA,TCA | A,S 302 | NP_001138552.2 | |
XM_005257236.3 | 1101 | Missense Mutation | GCA,TCA | A,S 302 | XP_005257293.1 |