Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTCCCTCCAGGAGCCGTCGGTA[C/T]GTGGTGATTTCCTTCTCCAGGTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606194 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT23 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRT23 - keratin 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282433.1 | 811 | Silent Mutation | ACA,ACG | T,T 233 | NP_001269362.1 | |
NM_015515.4 | 811 | Silent Mutation | ACA,ACG | T,T 370 | NP_056330.3 | |
XM_005257200.4 | 811 | Silent Mutation | ACA,ACG | T,T 233 | XP_005257257.1 | |
XM_011524595.1 | 811 | Silent Mutation | ACA,ACG | T,T 233 | XP_011522897.1 |