Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTAGATGGTTCTCATGCGTTAG[C/T]CATGGCCTGGCTACTAGGCCCTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611418 MIM: 611214 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SGSM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SGSM2 - small G protein signaling modulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098509.1 | Intron | NP_001091979.1 | ||||
NM_014853.2 | Intron | NP_055668.2 | ||||
XM_011524101.2 | Intron | XP_011522403.1 | ||||
XM_011524102.2 | Intron | XP_011522404.1 | ||||
XM_011524103.2 | Intron | XP_011522405.1 | ||||
XM_011524104.2 | Intron | XP_011522406.1 | ||||
XM_011524105.2 | Intron | XP_011522407.1 | ||||
XM_011524106.1 | Intron | XP_011522408.1 | ||||
XM_011524107.2 | Intron | XP_011522409.1 | ||||
XM_011524108.1 | Intron | XP_011522410.1 | ||||
XM_011524109.2 | Intron | XP_011522411.1 | ||||
XM_011524110.2 | Intron | XP_011522412.1 | ||||
XM_017025474.1 | Intron | XP_016880963.1 | ||||
XM_017025475.1 | Intron | XP_016880964.1 | ||||
XM_017025476.1 | Intron | XP_016880965.1 |
SNORD91A - small nucleolar RNA, C/D box 91A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSR1 - TSR1, ribosome maturation factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |