Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTGGAACACAGGCAGGACAAGG[A/G]TTCTGGAGGTTCAGGCCGGAAGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
QRICH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
QRICH2 - glutamine rich 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032134.2 | 9676 | Intron | NP_115510.1 | |||
XM_005257728.3 | 9676 | UTR 3 | XP_005257785.1 | |||
XM_006722136.3 | 9676 | UTR 3 | XP_006722199.1 | |||
XM_011525344.2 | 9676 | Intron | XP_011523646.1 | |||
XM_017025206.1 | 9676 | UTR 3 | XP_016880695.1 | |||
XM_017025207.1 | 9676 | Intron | XP_016880696.1 | |||
XM_017025208.1 | 9676 | UTR 3 | XP_016880697.1 |
UBALD2 - UBA like domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182565.3 | 9676 | Intron | NP_872371.1 |