Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGAAAGTTGGAAGAAGTACTGC[A/G]ACATCGTAGAAAGGAGTAGCTGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601025 MIM: 612128 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP2B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP2B1 - adaptor related protein complex 2 beta 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASL10B - RAS like family 10 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033315.3 | 102 | Intron | NP_201572.1 | |||
XM_017025300.1 | 102 | Intron | XP_016880789.1 | |||
XM_017025301.1 | 102 | UTR 5 | XP_016880790.1 |