Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGACTCTATAGCTGATTTTCTC[A/G]ATAGTGATGAGGAAGAAGACAGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604500 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYO19 - myosin XIX | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNHIT3 - zinc finger HIT-type containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281432.1 | 308 | Missense Mutation | AAT,GAT | N,D 79 | NP_001268361.1 | |
NM_001281433.1 | 308 | Intron | NP_001268362.1 | |||
NM_001281434.1 | 308 | Intron | NP_001268363.1 | |||
NM_004773.3 | 308 | Missense Mutation | AAT,GAT | N,D 79 | NP_004764.1 |