Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTCTGCACAGCAGCCACTGCCT[C/G]CTCCATCAGCGTCTCCAGCGCCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608588 MIM: 602301 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHX58 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHX58 - DEXH-box helicase 58 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024119.2 | 1732 | Missense Mutation | CAG,GAG | Q,E 505 | NP_077024.2 | |
XM_017025059.1 | 1732 | Missense Mutation | CAG,GAG | Q,E 505 | XP_016880548.1 | |
XM_017025060.1 | 1732 | Missense Mutation | CAG,GAG | Q,E 505 | XP_016880549.1 |
KAT2A - lysine acetyltransferase 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |