Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCCAGCACTTTCTGGAGGTCCG[C/T]CTTGGCCTCGGCCTCATTCCACACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604392 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AIPL1 - aryl hydrocarbon receptor interacting protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033054.2 | 876 | Missense Mutation | ACG,GCG | T,A 222 | NP_001028226.1 | |
NM_001033055.2 | 876 | Missense Mutation | ACG,GCG | T,A 225 | NP_001028227.1 | |
NM_001285399.2 | 876 | Missense Mutation | ACG,GCG | T,A 273 | NP_001272328.1 | |
NM_001285400.2 | 876 | Missense Mutation | ACG,GCG | T,A 263 | NP_001272329.1 | |
NM_001285401.2 | 876 | Missense Mutation | ACG,GCG | T,A 261 | NP_001272330.1 | |
NM_001285402.1 | 876 | Missense Mutation | ACG,GCG | T,A 246 | NP_001272331.1 | |
NM_001285403.2 | 876 | Intron | NP_001272332.1 | |||
NM_014336.4 | 876 | Missense Mutation | ACG,GCG | T,A 285 | NP_055151.3 |