Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCAAGAGCTGTAATATTTTTCT[C/T]CTGGGAAGCAATCTCTTCCATGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609106 MIM: 609002 | ||||||||||||||||||||
Literature Links: |
FBXO39 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO39 - F-box protein 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEKT1 - tektin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_053285.1 | 987 | Missense Mutation | AAG,GAG | K,E 293 | NP_444515.1 | |
XM_011524027.2 | 987 | Intron | XP_011522329.1 |