Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGACCATGGAGGAGCTGAAGGAC[C/G]TGCTGAAGGTGGGTGAGACGGGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604375 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL16 - ADP ribosylation factor like GTPase 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HGS - hepatocyte growth factor-regulated tyrosine kinase substrate | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004712.4 | 367 | Missense Mutation | CTG,GTG | L,V 95 | NP_004703.1 | |
XM_011525463.2 | 367 | Missense Mutation | CTG,GTG | L,V 95 | XP_011523765.1 | |
XM_017025297.1 | 367 | Missense Mutation | CTG,GTG | L,V 86 | XP_016880786.1 | |
XM_017025298.1 | 367 | UTR 5 | XP_016880787.1 |
MIR6786 - microRNA 6786 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |