Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCACCTTCAGATCAACCAGACAT[C/T]TGAGGAGCTGCGACTGGTCACGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601511 | ||||||||||||||||||||
Literature Links: |
STAT5A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STAT5A - signal transducer and activator of transcription 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288718.1 | 703 | Missense Mutation | TCT,TTT | S,F 148 | NP_001275647.1 | |
NM_001288719.1 | 703 | Missense Mutation | TCT,TTT | S,F 118 | NP_001275648.1 | |
NM_001288720.1 | 703 | Missense Mutation | TCT,TTT | S,F 148 | NP_001275649.1 | |
NM_003152.3 | 703 | Missense Mutation | TCT,TTT | S,F 148 | NP_003143.2 | |
XM_005257624.3 | 703 | Missense Mutation | TCT,TTT | S,F 148 | XP_005257681.1 |