Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAGCAATCTCCCTCCAGCTCTG[A/G]AGACAGGTGGGGCACAGCTTCCGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607273 MIM: 614960 | ||||||||||||||||||||
Literature Links: |
FLCN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLCN - folliculin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLD6 - phospholipase D family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178836.3 | 738 | Missense Mutation | TCC,TTC | S,F 231 | NP_849158.2 | |
XM_017024310.1 | 738 | Missense Mutation | CCA,TCA | P,S 230 | XP_016879799.1 |