Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCGCTGACCGGAGGTTCCATGG[A/G]CCCAGGGTAGGGCGGTGGTGGGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608897 MIM: 606962 | ||||||||||||||||||||
Literature Links: |
UNC13D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UNC13D - unc-13 homolog D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WBP2 - WW domain binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012478.3 | 516 | Missense Mutation | CCC,TCC | P,S 203 | NP_036610.2 | |
XM_011524580.2 | 516 | Missense Mutation | CCC,TCC | P,S 158 | XP_011522882.1 |