Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGAGACACCCCCAATCTGCCCT[C/T]AACCAGCCCAAGAAACCCTCTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614966 MIM: 608613 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC46 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC46 - leucine rich repeat containing 46 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCRN2 - secernin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SP6 - Sp6 transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258248.1 | 2353 | UTR 3 | NP_001245177.1 | |||
NM_199262.2 | 2353 | UTR 3 | NP_954871.1 | |||
XM_006722115.3 | 2353 | UTR 3 | XP_006722178.1 |