Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGAAAAAGCTCATATCAACATTC[A/G]AGATTCTATCCACCTCACACCAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606477 MIM: 611214 | ||||||||||||||||||||
Literature Links: |
SRR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SRR - serine racemase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304803.1 | 1348 | Intron | NP_001291732.1 | |||
NM_021947.2 | 1348 | Missense Mutation | CAA,CGA | Q,R 20 | NP_068766.1 | |
XM_006721565.3 | 1348 | Missense Mutation | CAA,CGA | Q,R 20 | XP_006721628.1 | |
XM_006721566.3 | 1348 | Missense Mutation | CAA,CGA | Q,R 20 | XP_006721629.1 | |
XM_011523974.2 | 1348 | Missense Mutation | CAA,CGA | Q,R 20 | XP_011522276.1 |
TSR1 - TSR1, ribosome maturation factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |