Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCGTGGTCCCTTTGAAGAGCTCCA[C/G]CAGCTCCTTGTAGTCCGGGTCGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616514 MIM: 176790 | ||||||||||||||||||||
Literature Links: |
MCRIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MCRIP1 - MAPK regulated corepressor interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001093767.2 | 530 | Intron | NP_001087236.1 | |||
NM_001288798.1 | 530 | Intron | NP_001275727.1 | |||
NM_001288799.1 | 530 | Intron | NP_001275728.1 | |||
NM_207368.4 | 530 | Intron | NP_997251.2 | |||
XM_011523571.2 | 530 | Intron | XP_011521873.1 | |||
XM_011523572.2 | 530 | Intron | XP_011521874.1 |
P4HB - prolyl 4-hydroxylase subunit beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R27 - protein phosphatase 1 regulatory subunit 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007533.3 | 530 | Missense Mutation | CTG,GTG | L,V 145 | NP_001007534.1 |