Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCGAGGGCAGGTTTGAGCCTCCC[A/G]CTGTCCCTGTCTCGAGTTATGTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602954 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAD51D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAD51D - RAD51 paralog D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142571.1 | 1087 | Silent Mutation | AGC,AGT | S,S 277 | NP_001136043.1 | |
NM_002878.3 | 1087 | Silent Mutation | AGC,AGT | S,S 257 | NP_002869.3 | |
NM_133629.2 | 1087 | Silent Mutation | AGC,AGT | S,S 145 | NP_598332.1 |
RAD51L3-RFFL - RAD51L3-RFFL readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |