Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTGCCCCACTGGGCCTTCTTCA[A/G]TCTGTGGAGACTTTAGGCTGAGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611508 MIM: 603060 | ||||||||||||||||||||
Literature Links: |
CAMTA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAMTA2 - calmodulin binding transcription activator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171166.1 | 828 | Intron | NP_001164637.1 | |||
NM_001171167.1 | 828 | Intron | NP_001164638.1 | |||
NM_001171168.1 | 828 | Intron | NP_001164639.1 | |||
NM_015099.3 | 828 | Intron | NP_055914.2 | |||
XM_006721478.3 | 828 | Intron | XP_006721541.1 | |||
XM_006721481.3 | 828 | Intron | XP_006721544.1 | |||
XM_006721482.3 | 828 | Intron | XP_006721545.1 | |||
XM_011523746.2 | 828 | Intron | XP_011522048.1 | |||
XM_011523747.2 | 828 | Intron | XP_011522049.1 | |||
XM_011523748.2 | 828 | Intron | XP_011522050.1 | |||
XM_011523749.2 | 828 | Intron | XP_011522051.1 |
INCA1 - inhibitor of CDK, cyclin A1 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167985.1 | 828 | Silent Mutation | CTG,TTG | L,L 118 | NP_001161457.1 | |
NM_001167986.1 | 828 | Silent Mutation | CTG,TTG | L,L 133 | NP_001161458.1 | |
NM_001167987.1 | 828 | Silent Mutation | CTG,TTG | L,L 133 | NP_001161459.1 | |
NM_213726.2 | 828 | Silent Mutation | CTG,TTG | L,L 118 | NP_998891.2 | |
XM_005256627.3 | 828 | Silent Mutation | CTG,TTG | L,L 133 | XP_005256684.1 | |
XM_005256628.4 | 828 | Silent Mutation | CTG,TTG | L,L 133 | XP_005256685.1 | |
XM_005256629.1 | 828 | Silent Mutation | CTG,TTG | L,L 118 | XP_005256686.1 | |
XM_006721517.1 | 828 | Silent Mutation | CTG,TTG | L,L 133 | XP_006721580.1 | |
XM_006721518.2 | 828 | Silent Mutation | CTG,TTG | L,L 118 | XP_006721581.1 | |
XM_006721519.3 | 828 | Silent Mutation | CTG,TTG | L,L 118 | XP_006721582.1 | |
XM_011523832.2 | 828 | Silent Mutation | CTG,TTG | L,L 133 | XP_011522134.1 | |
XM_011523834.2 | 828 | Missense Mutation | ACT,ATT | T,I 138 | XP_011522136.1 |
KIF1C - kinesin family member 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101927979 - translation initiation factor IF-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |