Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGTTCCAAAGTGGGCCTGGGCA[C/T]AGCCGAGGGCACCTCCACCCCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612341 MIM: 604885 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGT6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GGT6 - gamma-glutamyltransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122890.2 | 847 | Missense Mutation | NP_001116362.1 | |||
NM_001288702.1 | 847 | Missense Mutation | NP_001275631.1 | |||
NM_001288703.1 | 847 | Intron | NP_001275632.1 | |||
NM_001288704.1 | 847 | Intron | NP_001275633.1 | |||
NM_153338.3 | 847 | Missense Mutation | NP_699169.2 |
MYBBP1A - MYB binding protein 1a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |