Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGGACAGTGCATCCTCGTTCTC[C/T]GCTAGCTCAAACACCTCGATGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603325 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R9B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1R9B - protein phosphatase 1 regulatory subunit 9B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032595.4 | 1957 | Silent Mutation | GCA,GCG | A,A 651 | NP_115984.3 | |
XM_011525381.2 | 1957 | Intron | XP_011523683.1 |
SAMD14 - sterile alpha motif domain containing 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |