Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCCTATATAGATGTTGAGGTGC[A/G]GAGGCCGTGGGCTTTTGTTGGGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CYB5D2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYB5D2 - cytochrome b5 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001254755.1 | 295 | Intron | NP_001241684.1 | |||
NM_001254756.1 | 295 | Intron | NP_001241685.1 | |||
NM_144611.3 | 295 | Missense Mutation | AGA,GGA | R,G 5 | NP_653212.1 | |
XM_006721447.2 | 295 | Intron | XP_006721510.2 | |||
XM_011523649.2 | 295 | Intron | XP_011521951.1 | |||
XM_017024180.1 | 295 | Intron | XP_016879669.1 | |||
XM_017024181.1 | 295 | Intron | XP_016879670.1 | |||
XM_017024182.1 | 295 | Intron | XP_016879671.1 | |||
XM_017024183.1 | 295 | Missense Mutation | AGA,GGA | R,G 5 | XP_016879672.1 |
ZZEF1 - zinc finger ZZ-type and EF-hand domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015113.3 | 295 | Intron | NP_055928.3 | |||
XM_005256561.2 | 295 | Intron | XP_005256618.1 | |||
XM_011523759.2 | 295 | Intron | XP_011522061.1 | |||
XM_017024382.1 | 295 | Intron | XP_016879871.1 | |||
XM_017024383.1 | 295 | Intron | XP_016879872.1 | |||
XM_017024384.1 | 295 | Intron | XP_016879873.1 |