Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAAAGGCGTCGCCCGGGTCGCCG[C/T]CGCCGCCGCCGCTGCCGGCCCGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609878 MIM: 605550 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MED9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MED9 - mediator complex subunit 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASD1 - ras related dexamethasone induced 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199989.1 | 847 | UTR 3 | NP_001186918.1 | |||
NM_016084.4 | 847 | Missense Mutation | GAC,GGC | D,G 236 | NP_057168.1 |