Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGTGCTTCCGCAGCTGGGGGCTC[C/T]GCTCCATCTCCAGGGGATCCTCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608759 MIM: 610598 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYGB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYGB - cytoglobin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_134268.4 | 231 | Missense Mutation | CAG,CGG | Q,R 74 | NP_599030.1 | |
XM_005257005.2 | 231 | Missense Mutation | CAG,CGG | Q,R 74 | XP_005257062.1 | |
XM_017024116.1 | 231 | Missense Mutation | CAG,CGG | Q,R 9 | XP_016879605.1 |
PRCD - progressive rod-cone degeneration | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077620.2 | 231 | Intron | NP_001071088.1 | |||
XM_017025013.1 | 231 | Intron | XP_016880502.1 | |||
XM_017025014.1 | 231 | Intron | XP_016880503.1 | |||
XM_017025015.1 | 231 | Intron | XP_016880504.1 |