Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGATGGTGTACAGAGGGGAGCCT[A/G]GGATGGGTGCATCCTGAGTCACGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603892 | ||||||||||||||||||||
Literature Links: |
EFTUD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EFTUD2 - elongation factor Tu GTP binding domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142605.1 | 2989 | Missense Mutation | CCA,CTA | P,L 829 | NP_001136077.1 | |
NM_001258353.1 | 2989 | Missense Mutation | CCA,CTA | P,L 864 | NP_001245282.1 | |
NM_001258354.1 | 2989 | Missense Mutation | CCA,CTA | P,L 854 | NP_001245283.1 | |
NM_004247.3 | 2989 | Missense Mutation | CCA,CTA | P,L 864 | NP_004238.3 |
HIGD1B - HIG1 hypoxia inducible domain family member 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |