Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAGATGATGCCATATTGGGCCAG[C/T]TCGTGCAGCTCCAGGTGGTTGAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 180073 | ||||||||||||||||||||
Literature Links: |
PDE6G PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDE6G - phosphodiesterase 6G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002602.3 | 384 | Silent Mutation | GAA,GAG | E,E 80 | NP_002593.1 | |
XM_006721942.3 | 384 | Silent Mutation | GAA,GAG | E,E 130 | XP_006722005.1 | |
XM_017024734.1 | 384 | Silent Mutation | GAA,GAG | E,E 150 | XP_016880223.1 | |
XM_017024735.1 | 384 | Silent Mutation | GAA,GAG | E,E 80 | XP_016880224.1 | |
XM_017024736.1 | 384 | Silent Mutation | GAA,GAG | E,E 80 | XP_016880225.1 |
TSPAN10 - tetraspanin 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |