Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGACCGCGGTCTTGGAGCGCGCG[C/T]ACACCACGTGGTCGGGCGTGAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603198 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTRF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTRF - polymerase I and transcript release factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012232.5 | 1146 | Missense Mutation | TAC,TGC | Y,C 308 | NP_036364.2 | |
XM_005257242.3 | 1146 | Intron | XP_005257299.1 |