Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCACGAGTGACCCTGCTGCCCA[C/T]GCCCCACGTCAGCGGGCTGAGCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609046 | ||||||||||||||||||||
Literature Links: |
BTBD17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BTBD17 - BTB domain containing 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPR142 - G protein-coupled receptor 142 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181790.1 | 389 | Missense Mutation | ACG,ATG | T,M 130 | NP_861455.1 | |
XM_005257305.3 | 389 | Missense Mutation | ACG,ATG | T,M 42 | XP_005257362.1 |